Learn More
Thermo Scientific™ Dry-Bags™ Maximum Recovery Diluent (MRD)
Simplify media preparation in food laboratories with this high quality dehydrated medium in a convienient bag format; just open-fill-dispense, saving precious time in the lab.
Brand:  Thermo Scientific™ DB0733M
1215.44 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 12660299
  • £1270.00 / Each
Explore more special offers
Description
Show More
Specifications
Specifications
Clear, colorless
Dry-Bags Maximum Recovery Diluent (MRD)
Dehydrated Culture Media
20L
10°C to 30°C
Dry-Bags™, with filter
10/Pk.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
3 results found
Lot Number Certificate Type Date Catalog Number
Lot Number4460211 Certificate TypeCertificate of Analysis Date02/08/2024 Catalog NumberDB0733M
Lot Number4460102 Certificate TypeCertificate of Analysis Date02/08/2024 Catalog NumberDB0733M
Lot Number4379526 Certificate TypeCertificate of Analysis Date07/02/2023 Catalog NumberDB0733M
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Dry-Bags™ Maximum Recovery Diluent (MRD) >