Learn More
DWK Life Sciences DURAN™ Baffled Flask, with GL 45 thread
Due to the baffles incorporated in the product, a solution to be shaken breaks on it and oxygen is introduced into the solution.
Brand: DWK Life Sciences 212835459
33.05 GBP valid until 2025-06-13
Use promo code "24123" to get your promotional price.
Description
- Four baffled-bottom flasks cause a turbulent flow, increase the gas exchange surface and produce a higher oxygen transfer
- At higher shaking speeds, the baffles increase aeration for better growth, especially with bacteria
- The moulded bottom-baffles provide increased movement at lower speeds for a more gentle medium aeration of sensitive cultures
- The flasks can therefore be sealed with the tried-and-tested membrane screw cap from the DWK Life Sciences which enables a reproducible gas exchange compared to other sealing mechanisms e.g. sealing with cotton wool
- The flasks are manufactured in a fully automated process to provide totally consistent flask shapes leading to highly reproducible experimental conditions and results
- The special production process enables the manufacture of the product complete with thread in a two-stage process
- Available in range of three sizes for process scale up
- The increased wall thickness of the flasks ensures excellent mechanical stability and a long service life.
- Easy-to-read scale with graduation marks to estimate the content

Specifications
1000 mL | |
Growth and cultivation of microorganisms; Media preparation and cultivation; Storage and transport of bacterial, yeast, plant and animal cells; Suitable for liquid cultures (e.g. pure cultures, enrichment cultures); Use on shaker equipment; GL 45 necks allowing easy use with : GL 45 membrane screw cap for use during autoclaving, 30 mm culture plugs or stoppers for culturing and GL 45 screw caps for media or reagent storage | |
4/Pk. | |
Autoclavable |
Borosilicate Glass (3.3) | |
Membrane Cap and Pouring Ring | |
GL 45 |
Product Suggestions
Customers who viewed this item also viewed.
Videos
-
Baffled flask
Special Offers
-
Multiple Manufacturers
Back to the Lab Summer Semester
Save up to 94% on Teaching Lab EssentialsPromo Code: 25721Expires: 2025-05-01
New Offer Save Now -
Multiple Manufacturers
Price-Shattering Deals on Lab Glassware
Stock up on lab beakers, flasks and more before these deals disappear.Promo Code: 24123Expires: 2025-06-14
New Offer Save Now
Your input is important to us. Please complete this form to provide feedback related to the content on this product.