Learn More
DWK Life Sciences DURAN™ Pipeline Filter
A pipeline filter is used to separate solids from a gas phase.
£55.55 - £228.00
Specifications
Material | Borosilicate Glass (3.3) |
---|---|
For Use With (Application) | In-line filtration of gas lines to remove solid impurities (e.g. dust); Qualitative inorganic analysis and preparative chemistry; Filtration at reduced pressure; Filtering |
No. per Pack | 1/Pk. |
Product Code | Brand | Diameter (Metric)Top | Description | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric)Top | Description | Price | Quantity & Availability | ||||
13151554
|
DWK Life Sciences
258551109 |
60 mm plate | Porosity 1 |
£126.00
Each |
Estimated Shipment: 16-08-2024
Log in to see stock. |
||||
10273361
|
DWK Life Sciences
258551203 |
60 mm plate | Porosity 2 |
£126.00
Each |
Estimated Shipment: 13-09-2024
Log in to see stock. |
||||
10018210
|
DWK Life Sciences
258551306 |
60 mm plate | Porosity 3 |
£126.00
Each |
Estimated Shipment: 13-09-2024
Log in to see stock. |
||||
10484261
|
DWK Life Sciences
258551409 |
60 mm plate | Porosity 4 |
£137.00
Each |
Estimated Shipment: 13-09-2024
Log in to see stock. |
||||
10067940
|
DWK Life Sciences
258550104 |
30 mm | Porosity 1 |
£55.55
Each |
Estimated Shipment: 13-09-2024
Log in to see stock. |
||||
10077940
|
DWK Life Sciences
258550207 |
30 mm | Porosity 2 |
£80.25
Each |
Estimated Shipment: 16-08-2024
Log in to see stock. |
||||
10539061
|
DWK Life Sciences
258550301 |
30 mm | Porosity 3 |
£55.55
Each |
Estimated Shipment: 13-09-2024
Log in to see stock. |
||||
10710812
|
DWK Life Sciences
258552105 |
90 mm | Porosity 1 |
£228.00
Each |
Estimated Shipment: 13-09-2024
Log in to see stock. |
||||
10131891
|
DWK Life Sciences
258550404 |
30 mm | Porosity 4 |
£60.70
Each |
Estimated Shipment: 13-09-2024
Log in to see stock. |
||||
10325111
|
DWK Life Sciences
258552208 |
90 mm | Porosity 2 |
£228.00
Each |
Estimated Shipment: 13-09-2024
Log in to see stock. |
||||
10078030
|
DWK Life Sciences
258552302 |
90 mm | Porosity 3 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
In addition to solid-liquid filtration, filtration or separation of reaction mixtures from the gas phase is also possible. If, for example, a pipeline filter is installed in a hose system, these filters solid residues from the gas phase and thus purifies it.
- By installing the filter in a pipe system, the purification of a carrier gas is possible
- Fired-on, highly durable white print
Specifications
Borosilicate Glass (3.3) | |
In-line filtration of gas lines to remove solid impurities (e.g. dust); Qualitative inorganic analysis and preparative chemistry; Filtration at reduced pressure; Filtering | |
1/Pk. |