missing translation for 'onlineSavingsMsg'
Learn More
DWK Life Sciences DURAN™ PTFE-coated Silicone Septum Seal, Platinum cured silicone, Septum for Piercing, Use with Open Topped Screw Cap Product Code.: 15738358
Add to basket

DWK Life Sciences DURAN™ PTFE-coated Silicone Septum Seal, Platinum cured silicone, Septum for Piercing, Use with Open Topped Screw Cap

Product Code. 15738358 Shop All DWK Life Sciences Products
GL 45
Click to view available options
GL 45
This item is not returnable. View return policy

Product Code. 15738358

Brand: DWK Life Sciences 292483005

Add to basket
Add to basket
This item is not returnable. View return policy

High purity PTFE coated silicone septum

High purity PTFE coated silicone septum for use with DURAN™ GL 45 open topped screw cap and all sizes of DURAN™ GL 45 laboratory bottles. Can be used for the addition, inoculation or sampling using a syringe and needle. Ideal for chemistry, life science and biopharma laboratories.

  • Soft silicone rubber for easy needle penetration and resealability
  • Manufactured from high purity, Platinum catalysed silicone rubber and high quality PTFE film, reducing the risk of solvent contamination
  • The silicone is faced with highly resistant PTFE to protect the silicone from chemical attack by the solvents
  • The septum is suitable for use with all GL 45 sizes of the DURAN™ GL 45 bottles from 100 to 20,000 ml
  • A liquid tight seal is provided when used in combination with the red PBT GL 45 open topped screw cap
  • Wide temperature usage range from – 50°C up to + 180°C
  • Autoclavable at 121°C for 15 minutes
  • Dry heat sterilizable at up to 180°C
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Autoclaving of media and reagents; Preparation of media and reagents; Chemical and thermal reactions; Sample storage; Storage of media and reagents
Material Silicone, PTFE
Quantity 10/Pk.
Thickness (Metric) 3 mm
Thread Size (Metric) GL 45
Autoclavable Not Autoclavable
Diameter (Metric) 43 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
DWK Life Sciences DURAN™ PTFE-coated Silicone Septum Seal, Platinum cured silicone, Septum for Piercing, Use with Open Topped Screw Cap >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.