missing translation for 'onlineSavingsMsg'
Learn More
DWK Life Sciences DURAN™ Reagent Bottle, Wide Neck, Clear, Without Stopper Product Code.: 11706263
Estimated Shipment: 28-07-2025
to see stock.
Add to basket

DWK Life Sciences DURAN™ Reagent Bottle, Wide Neck, Clear, Without Stopper

Product Code. 11706263 Shop All DWK Life Sciences Products
100 mL
Click to view available options
Capacity (Metric):
10,000 mL
100 mL
1000 mL
20,000 mL
2000 mL
250 mL
50 mL
500 mL
5000 mL
This item is not returnable. View return policy

Product Code. 11706263

Brand: DWK Life Sciences 211842402

Estimated Shipment: 28-07-2025
to see stock.
Add to basket
Add to basket
This item is not returnable. View return policy

Classic wide-neck, glass-stoppered reagent bottles.

The classical stoppered reagent bottles are moulded from DURAN™ borosilicate 3.3 Type 1 glass, and are suitable for the storage of liquid and solid chemicals and reagents. The bottles feature a wide conical ground joint neck and comply with ISO 4796-2. The wider neck is more convenient for handling powders. The glass stopper provides an excellent seal that protects the contents from evaporation, oxidation, and contamination. The bottles have a Retrace Code for manufacturing lot traceability and a marking field in permanent white enamel.

  • Easy access to the bottle contents due to the wide diameter of the bottle neck
  • All borosilicate glass components and therefore suitable for storage of aggressive materials
  • Tight closure due to standard ground joint
  • Easy-to-read scale with graduation marks to estimate the content
  • Fired-on, highly durable white ceramic print
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 100 mL
Description Neck with standard ground joint
For Use With (Application) Storage of air sensitive reagents, pharmaceuticals, and chemicals; Storage of hygroscopic liquids and pharmaceuticals; Transport and shipping of samples
Material Borosilicate Glass (3.3)
Quantity 10/Pk.
Autoclavable Autoclavable
Diameter (Metric) 52 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
DWK Life Sciences DURAN™ Reagent Bottle, Wide Neck, Clear, Without Stopper >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.