missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences DURAN™ YOUTILITY™ Label, Printable self-adhesive
Shop All DWK Life Sciences Products
Click to view available options
White polyester
Description
The accurate and reliable labelling of scientific samples is an integral part of the success of your laboratory. Labels carry critical information, and without accurate labelling, months of research could be put at risk. Identification labels are the foundation of an effective laboratory management process.
- High temperature performance range makes them ideal for use in fridges, freezers, autoclaves, incubators, and water baths
- Can be printed with office printers, copiers, or marked with technical lab pens
- The labels have a high adhesive strength they are peelable and easy to remove which safes time for cleaning
- An free easy-to-use label creator web app for individual label design is available
- Chemically resistant to many typical laboratory chemicals, and solvents
- The labels are non-hazardous and free of toxic heavy metals

Specifications
Specifications
Color | White |
For Use With (Application) | Labelling on YOUTILITY bottles; Sample labelling |
Material | Polyester |
Quantity | 1 x 100 |
Autoclavable | Autoclavable |
Special Offers
-
Multiple Manufacturers
Chromatography and Chemicals Promotion
Explore our new promotion and get up to 70% off some of our most popular products.Promo Code: 24127Expires: 2025-05-31
New Offer Save Up to 70%
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
DWK Life Sciences DURAN™ YOUTILITY™ Label, Printable self-adhesive >
Spot an opportunity for improvement?Share a Content Correction