missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ E-Gel™ Double Comb Agarose Gels with SYBR™ Safe DNA Gel Stain
GREENER_CHOICE

Invitrogen E-Gel Double Comb Agarose Gels with SYBR Safe DNA Gel Stain are designed for fast, convenient, and less-hazardous DNA sample electrophoresis.

Brand:  Invitrogen™ A42348

 View more versions of this product

Product Code. 16618485

  • £194.00 / Pack of 10

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

E-Gel™ 1 Kb Plus Express Ladder
2%
Agarose Gel
SYBR™ Safe
Less hazardous, less waste, fewer resources used
Double Comb
• Each box contains 10 gels

Store strictly at room temperature. Do not freeze.
Avoid cold spots near freezers or other uncontrolled environments.
E-Gel
50 to 2000 bp
22-well
10 Gels
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberYS130125-01 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog NumberA42348
Lot NumberYS050924-01 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog NumberA42348
Lot NumberYS141024-01 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog NumberA42348
Lot NumberYS121124-01 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog NumberA42348
Lot NumberYS200322-01 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberA42348
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ E-Gel™ Double Comb Agarose Gels with SYBR™ Safe DNA Gel Stain > Double Comb Agarose Gels with SYBR™ Safe DNA Gel Stain, 2%, 10 Gels

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.