Learn More
Invitrogen™ Silencer™ Human Genome siRNA Library
The Silencer™ Human Genome siRNA Library contains a total of 65,058 siRNAs targeting 21,686 genes extracted from the NCBI RefSeq database.
Brand: Invitrogen™ A30076
Description
Epigenetic regulation of gene expression is proving to be a key regulator in the development and progression of a host of human diseases. The Silencer™ Human Epigenetics siRNA Library contains 1,560 Ambion™ Silencer siRNAs targeting 520 epigenetic enzymes, including those mediating epigenetic signaling through the acetylation and methylation of histones. The library was designed using the most up-to-date genome databases, including the NCBI RefSeq database and cross-referenced to the Gene Ontology Consortium (GO) database and/or the HUGO Gene Nomenclature Committee (HGNC). Utilizing Ambion Silencer technology built on stringent bioinformatics to ensure high target specificity and siRNA potency, the Silencer Human Epigenetics siRNA Library is an ideal RNAi screening solution for focused and cost-effective epigenetic gene knockdown studies
Why use the Silencer Human Genome siRNA Library?Thermo Fisher Scientific Silencer siRNAs are optimized for design and high quality synthesis with guaranteed silencing (learn more about the guarantee). The Silencer Human Genome siRNA Library contains pre-designed and when available, validated siRNAs generated with potency and specificity in mind—so you can focus on data quality and not performance issues (learn more about the design). The library is delivered with full siRNA sequence information, as well as the exon targeted, each targeted gene's symbol, full name, aliases, RefSeq number, and Locus ID; plus results from any validation experiments performed using the included siRNAs. Please contact us for the complete list of gene targets.Silencer siRNA libraries allow you to screen efficiently and maximize data outputInvitrogen Silencer siRNAs are designed with potency in mind to help ensure gene knockdown at minimum concentrations. This not only helps ensure less potential for off target effects and false positive data but also minimizes false negative outcomes of siRNA screening. The Silencer Human Genome siRNA Library contains 0.25 nmol of each siRNA, sufficient for 100 transfections (at 25 nM siRNA, 100 μL transfection volume).Flexibility to meet your workflow needsThermo Fisher Scientific offers a comprehensive list of pre-defined or custom siRNA libraries to any set of human, mouse or rat genes with 1, 2 or 5 nmol of each siRNA or larger sizes. For a complete gene list, additional information or to discuss your research with one of our technical experts, contact your local sales representative or e-mail us at RNAilibraries@lifetech.com.- Comprehensive—Accelerate siRNA screening with complete and ready-to-ship collections of Invitrogen Silencer human genome siRNAs
- Reliable—Increase target screening accuracy with three individual siRNAs per gene target
- Effective—Reduce off target impacts at low concentrations with proven Invitrogen Silencer siRNA design
- Convenient—Reduce data de-convolution and accelerate target validation with our arrayed format
- Customizable—Choose the best option available to meet your workflow demands, including additional siRNA scales and pooled formats
Specifications
65,058 total siRNAs targeting 21,686 genes plated onto 384-well plates (186 plates) |
Product Suggestions
Customers who viewed this item also viewed.
For research use only. Not for use in diagnostic procedures