missing translation for 'onlineSavingsMsg'
Learn More

Novex™ Endogenous Biotin-Blocking Kit

Endogenous Biotin-Blocking Kit

Brand:  Novex™ E21390

Product Code. 10194472

  • £244.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 
This item is not returnable. View return policy

Description

Description

The Endogenous Biotin-Blocking Kit provides a method and reagents for a pretreatment that reduces or eliminates background signals when biotin-avidin or biotin-streptavidin detection systems are used to identify cellular targets.

The Endogenous Biotin-Blocking Kit offers:
  • Improved results—increased signal-to-noise ratio facilitates cell target identification
  • Ease of use—pipette-free dropper bottle offers speed and convenience
  • Fast—endogenous background reduction in under an hour
  • Flexibility—the kit can be used to help improve results from conventional immunohistochemistry, immunocytochemistry, RNA FISH, and ELISA procedures

    Biotin occurs naturally, functioning as an enzyme cofactor in the cytosol and mitochondria of a wide variety of cell types. The Endogenous Biotin-Blocking Kit minimizes interference from the endogenous biotin found in enzymes of mammalian cells and tissues, through a two-step blocking strategy. First, an excess of unlabeled streptavidin is added to the specimen to bind endogenous biotin-rich enzymes. The streptavidin is subsequently blocked with an excess of unlabeled biotin, effectively rendering the cell or tissue sample free of available biotin-binding sites.
    TRUSTED_SUSTAINABILITY
  • Specifications

    Specifications

    Endogenous Biotin-Blocking Kit
    Store in refrigerator at 2°C to 8°C
    Room Temperature
    Blocking⁄Background Suppression Reagent
    1 kit
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Manuals

    Certificates

    Certificates

    A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

    5 results found
    Lot Number Certificate Type Date Catalog Number
    Lot Number2856048 Certificate TypeCertificate of Analysis Date07/05/2024 Catalog NumberE21390
    Lot Number2610308 Certificate TypeCertificate of Analysis Date10/07/2023 Catalog NumberE21390
    Lot Number570745 Certificate TypeCertificate of Analysis Date06/10/2022 Catalog NumberE21390
    Lot Number2506003 Certificate TypeCertificate of Analysis Date12/09/2022 Catalog NumberE21390
    Lot Number2431313 Certificate TypeCertificate of Analysis Date20/01/2022 Catalog NumberE21390
    Special Offers

    Special Offers

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title
    Novex™ Endogenous Biotin-Blocking Kit > 1 kit

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

    For Research Use Only. Not for use in diagnostic procedures.