missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Vertical Gel Tank

Incorporates a sealing system which is compatible with all major types of 80mm x 100mm and 100mm x 100mm precast gel. Fisherbrand™ Vertical Gel Tank's ultra soft silicone seals and pressure bars which surround the glass plates guarantee leak proof gel casting.

Brand:  Fisherbrand™ FCVS10CBS

 View more versions of this product

Product Code. 11883293

  • ÂŁ1066.00 / Each
Estimated Shipment: 09-05-2025
to see stock.
Explore more special offers
Add to basket

Description

Description

Incorporates a sealing system which is compatible with all major types of 80mm x 100mm and 100mm x 100mm precast gel. Gel casting and running utilise the same insert, no transfer of glass plates during gel casting is necessary. The insert contains sliding clamps allowing very rapid set up of both hand cast and precast gels. Ultra soft silicone seals and pressure bars which surround the glass plates guarantee leak proof gel casting. 2mm thick glass plates prevent breakage and have bonded spacers for convenience. Accessory electroblotting and tube gel modules are available which use the same outer tank and lid.
  • Injection molded construction, durable and leakproof
  • Compatible with all 80mm x 100mm and 100mm x 100mm precast gels
  • Low buffer volumes
  • Run up to four gels
  • Interchangable modules for IEF/2D electrophoresis and electroblotting in a universal tank For Mini SDS PAGE, native PAGE, gradient, second dimension and nucleic acid separations.
Specifications

Specifications

80 mm x 100 mm and 100 mm x 100 mm PreCast Gels
1 Set Verti-Gel mini Complete System for Electrophoresis and Blotting
2.6 kg
Product Suggestions

Product Suggestions

Videos

Videos

  • Fisherbrand and Fisher Bioreagents - Focus on electrophoresis
    Fisherbrand provides a comprehensive range of horizontal gel systems for all your nucleic acid gel electrophoresis needs.
    Video Player is loading.
    Current Time 0:00
    Duration 0:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:00
     
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected

      Fisherbrand and Fisher BioReagents Focus on electrophoresis

    • Fisherbrand - Meet the Family
      Fisher Scientific's private label product line, Fisherbrand™, offers customers a broad selection of laboratory supplies & consumables, covering a range of applications such as pH & electrochemistry, stirring & mixing, centrifugation, liquid handling,
      Video Player is loading.
      Current Time 0:00
      Duration 0:00
      Loaded: 0%
      Stream Type LIVE
      Remaining Time 0:00
       
      1x
      • Chapters
      • descriptions off, selected
      • captions off, selected

        Fisherbrand Meet the Family

      SDS
      Documents

      Documents

      Certificates
      View accessory products
      Special Offers

      Special Offers

      Product Content Correction

      Your input is important to us. Please complete this form to provide feedback related to the content on this product.

      Product Title
      Fisherbrand™ Vertical Gel Tank > Weight: 2.6kg

      By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

      Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.