Learn More
Sartorius Entris™ II Precision Balance, Internal Adjustment
Entris™ II Precision Balance with Internal Adjustment offering isoCAL, LED touch technology, and 12 built-in applications at a budget price. Every balance offers quality, value and consistency.
Brand:  Sartorius BCE3202I-1CEU
1726.45 GBP valid until 2025-12-31
Use promo code "24377" to get your promotional price.
 View more versions of this product
Product Code. 16334708
  • £2120.00 / 1 unit
Explore more special offers
Description
Description
  • No matter what you're weighing, the new Entris™ II balance is always the right choice.
  • Offering unrivalled value and backed by almost 150 years of German engineering expertise, the Entris™ II comes in two product lines, so you can find the balance that meets your specific weighing needs.
  • With over 40 models from which to choose, you will surely find a model to meet your basic weighing needs.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CEU
3200 g
10 mg
CEU Electronic Toploading Balance
Precision Balance
RS232, USB C
Touchscreen
PBT, Glass, Stainless Steel
≤1.0 sec.
100 – 240 VAC (±10%), 50 - 60 Hz, 0.2 A
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products
Special Offers
Special Offers
Product Content Correction
Product Title
Sartorius Entris™ II Precision Balance, Internal Adjustment > Entris™ II Precision Balance, Internal Adjustment; Capacity: 3200 g; Readability: 10 mg; Model: CEU Electronic Toploading Balance