Learn More
Fisherbrand™ EPA Screw Neck Vial, Glass
Broad range of EPA Vials in clear and amber glass.
£49.84 - £172.85
Specifications
Description Screw Neck
Diameter (Metric) Outer 27.5 mm
Material Glass
For Use With (Application) TOC (EPA)
Thread Style 24-400
View More Specs
Products 8
Product Code Brand Volume (Metric) Color Price Quantity & Availability  
Product Code Brand Volume (Metric) Color Price Quantity & Availability  
10465982
View Documents Promotion Details
Fisherbrand™
24 09 0402
40 mL Clear
£76.75
Pack of 100
In Stock
to see stock.
Edge
Add to basket
10000782
View accessory products View Documents Promotion Details
Fisherbrand™
24 09 0589
20 mL Clear
£49.84
Pack of 100
In Stock
to see stock.
Edge
Add to basket
10758874
View Documents Promotion Details
Fisherbrand™
24 09 0839
30 mL Clear
£77.00
Pack of 100
In Stock
to see stock.
Edge
Add to basket
10458082
View Documents Promotion Details
Fisherbrand™
24 09 0927
20 mL Amber
£128.00
Pack of 100
In Stock
to see stock.
Edge
Add to basket
11510585
View Documents Promotion Details
Fisherbrand™
24 09 0923
30 mL Amber
£119.00
Pack of 100
In Stock
to see stock.
Add to basket
11530585
View Documents Promotion Details
Fisherbrand™
24 09 0928
40 mL Amber
£107.12
Pack of 100
In Stock
to see stock.
Edge
Add to basket
11540585
View Documents Promotion Details
Fisherbrand™
24 09 1089
60 mL Clear
£137.77
Pack of 100
In Stock
to see stock.
Edge
Add to basket
11550585
View Documents Promotion Details
Fisherbrand™
24 09 1090
60 mL Amber
£172.85
Pack of 100
In Stock
to see stock.
Add to basket
Description
Description
  • EPA Vials can be delivered with a certificate of cleanliness
  • EPA Vials can be obtained with any type of Screw Seal ND24 already screwed on
  • Clear and amber glass
  • Available in 20mL, 30mL, 40mL and 60mL
Specifications
Specifications
Screw Neck
Glass
24-400
27.5 mm
TOC (EPA)
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Fisherbrand™ EPA Screw Neck Vial, Glass