missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Epoxy-activated Sepharose™ 6B Medium

A pre-activated medium for immobilization of various ligands including sugars through coupling of hydroxy, amino or thiol groups on the ligand to Sepharose 6B via a 12-atom hydrophilic spacer arm

Brand:  Cytiva 17-0480-01

Product Code. 11534915

  • £550.00 / 15g

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Group to be coupled: -NH2, -OH, or -SH
  • Matrix: Agarose, 6%
  • Average particle size: 90μm
  • Spacer arm: hydrophilic 12 atom

TRUSTED_SUSTAINABILITY
Specifications

Specifications

6% Agarose
2 to 14
2°C to 8°C
90 μm
15 g
For immobilization of various ligands including sugars through coupling of Hydroxy, Amino or Thiol groups on the ligand to Sepharose 6B via a 12-atom hydrophilic spacer arm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cytiva Epoxy-activated Sepharose™ 6B Medium > Epoxy-activated Sepharose 6B

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.