Learn More
Eppendorf™ S-4-72 Rotor Adapters
Available in a variety of configurations to suite a wide range of applications.
Brand: Eppendorf™ 5804792009
190.00 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- For use with Centrifuge 5804/5804 R and 5810/5810 R
Specifications
Adapter | |
12 x 7/17 mL Tubes | |
S-4-72 (4 x 250mL) rotor in Model 5804/5810 centrifuges | |
7/17 mL |
Product Content Correction
Product Title
Eppendorf™ S-4-72 Rotor Adapters > For 12 x 7/17mL tubes
Spot an opportunity for improvement?Share a Content Correction