Learn More
Thermo Scientific™ Deionizer Cartridges
Choose from an array of Thermo Scientific™ Deionizer Cartridges for select Thermo Scientific™ Barnstead™ water systems, based on your application.
Brand:  Thermo Scientific™ D0760
 View more versions of this product
Product Code. 12740666
  • £273.00 / Each
Estimated Shipment: 25-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Choose from an array of Thermo Scientific™ Deionizer Cartridges based on your application needs. These cartridges are for use with the Thermo Scientific™ Barnstead™ Bantam™, B-Pure™ or Nanopure™ water purification systems as indicated.
  • Choice of full or half cartridges
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cartridge for anion removal; strong anion exchange; full size
Strong Anion Exchange
Bantam™ Deionizer, B-Pure™
1680 grains
Anion Removal
Deionizer Cartridge
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Deionizer Cartridges > Cartridge for anion removal; strong anion exchange; full size