Learn More
Eppendorf™ Stackable PCR Racks

For handling and storing tubes and plates
Brand: Eppendorf™ 0030124545
140.60 GBP valid until 2025-04-30
Use promo code "24138" to get your promotional price.
Description
- Compatible for 0.1, 0.2 and 0.5mL tubes and 96-well plates
- Stackable

Specifications
Assorted | |
Microtubes (0.1, 0.2, and 0.5 mL) | |
10/Pk. |
Product Suggestions
Customers who viewed this item also viewed.
Special Offers
-
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
Your input is important to us. Please complete this form to provide feedback related to the content on this product.