missing translation for 'onlineSavingsMsg'
Learn More
Ethyl 2-amino-4-(2-thienyl)thiophene-3-carboxylate, 97%, Thermo Scientific™ Product Code.: 15494516

Ethyl 2-amino-4-(2-thienyl)thiophene-3-carboxylate, 97%, Thermo Scientific™

Product Code. 15494516
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 15494516

Brand: Thermo Scientific Alfa Aesar H58777.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 243669-48-9
Molecular Formula C11H11NO2S2
Molecular Weight (g/mol) 253.33
MDL Number MFCD00115120
InChI Key AKFDBWIUWIWHRK-UHFFFAOYSA-N
Synonym ethyl 2-amino-4-2-thienyl thiophene-3-carboxylate, ethyl 5'-amino-2,3'-bithiophene-4'-carboxylate, ethyl 2-amino-4-thiophen-2-yl thiophene-3-carboxylate, 5'-amino-2,3' bithiophenyl-4'-carboxylic acid ethyl ester, ethyl 5'-amino 2,3'-bithiophene-4'-carboxylate, ethyl 2-azanyl-4-thiophen-2-yl-thiophene-3-carboxylate, 2,3'-bithiophene-4'-carboxylicacid, 5'-amino-, ethyl ester, 2-amino-4-thiophen-2-yl-3-thiophenecarboxylic acid ethyl ester, maybridge1_003774Show MoreShow Less
PubChem CID 718227
SMILES CCOC(=O)C1=C(N)SC=C1C1=CC=CS1

Specifications

Melting Point 85°C to 87°C
Quantity 5 g
IUPAC Name ethyl 5'-amino-[2,3'-bithiophene]-4'-carboxylate
Formula Weight 253.33
Percent Purity 97%
Chemical Name or Material Ethyl 2-amino-4-(2-thienyl)thiophene-3-carboxylate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ethyl 2-amino-4-(2-thienyl)thiophene-3-carboxylate, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.