missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 3-exo-aminobicyclo[2.2.1]hept-5-ene-2-exo-carboxylate hydrochloride, 99%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass Bottle
Specifications
Specifications
Melting Point | 129°C to 132°C |
Color | White to Beige |
Infrared Spectrum | Authentic |
Assay Percent Range | 98.5% min. (Argentometry) |
Packaging | Glass Bottle |
Solubility Information | Solubility in water: soluble. Other solubilities: soluble in ethanol |
Quantity | 1 g |
Formula Weight | 217.69 |
Percent Purity | 99% |
Physical Form | Fine Crystalline Powder |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 10
Safety and Handling
Product Identifier
- Ethyl 3-exo-aminobicyclo¢2.2.1!hept-5-ene-2-exo-carboxylate hydrochloride
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Ethyl 3-exo-aminobicyclo[2.2.1]hept-5-ene-2-exo-carboxylate hydrochloride, 99%, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction