missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MULTIROIR Euronorm ESD Container
Simplified cleaning thanks to smooth internal walls designed for the storage and transport of electronic products
Brand: MULTIROIR AESDEG64/42HG
This item is not returnable.
View return policy
Description
- Euronorm ESD container
- Material: polypropylene 2 integrated ergonomic handles
- Simplified cleaning thanks to smooth internal walls
- Specially designed for the storage and transport of electronic products
Specifications
Storage Box | |
Gray | |
BP6420 K |
Polypropylene | |
Rectangular | |
600 x 400 x 420 mm |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
MULTIROIR Euronorm ESD Container > 600 x 400 x 420 mm
Spot an opportunity for improvement?Share a Content Correction