missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Temperature Probe Accessory for FB705 Sonicator

For monitoring temperature of sensitive samples

Brand:  Fisherbrand™ FB4102

 View more versions of this product

Product Code. 12911161

  • £249.00 / Each
Estimated Shipment: 24-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • For monitoring temperature sensitive samples volumes
  • Can be used to monitor volumes from 100 - 1000mL
  • 6 in. long Temperature monitoring probe (Stainless Steel)
Specifications

Specifications

Temperature Probe
15.24 cm
FB 705 Sonic Dismembrator™
6 in.
Ideal for monitoring temperature of samples 100mL or larger
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisherbrand™ Temperature Probe Accessory for FB705 Sonicator > Temperature Probe

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.