Learn More
Fisherbrand™ BloodBloc™ Biohazard Wipes, Liners and Rolls
Prevent leakage of fluids and chemicals to hands, gloves and work surfaces
Brand: Fisherbrand™ 0667050
Description
Wipes and Liners are non-sterile and not intended for use on patients.
- Soft, white absorbent wipe with bright-orange polyethylene backing is impermeable to blood, body fluids and strong solvents including xylene, xylene substitutes, acetones, ketones, alcohols and formalin
- Nonwoven cellulose captures and retains fluid; nonlinting for use with delicate instrumentation such as blood counters
- Orange flexible impermeable plastic backing prevents soak-through of pathogens and chemicals to gloves which help minimize cross-contamination when lab personal open doors or touch surfaces
- Can also be used as mats and liners; rolls can be cut to desired length
- One time use only
Specifications
Biohazard Wipe | |
10.16 cm | |
10.16 cm | |
Orange backing | |
4 x 4 in. (10.16 x 10.16 cm) | |
Blood tubes, microbiology plates, pipettes, probes, specimen containers, instruments |
4 in. | |
4 in. | |
Heavy | |
Single-use | |
Prevent soak through of blood, body fluids, tissue and chemicals to gloves and surfaces when handling tubes, specimens, plates. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.