missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ BloodBloc™ Biohazard Wipes, Liners and Rolls
Prevent leakage of fluids and chemicals to hands, gloves and work surfaces
Brand: Fisherbrand™ 0667051
This item is not returnable.
View return policy
Description
Wipes and Liners are non-sterile and not intended for use on patients.
- Soft, white absorbent wipe with bright-orange polyethylene backing is impermeable to blood, body fluids and strong solvents including xylene, xylene substitutes, acetones, ketones, alcohols and formalin
- Nonwoven cellulose captures and retains fluid; nonlinting for use with delicate instrumentation such as blood counters
- Orange flexible impermeable plastic backing prevents soak-through of pathogens and chemicals to gloves which help minimize cross-contamination when lab personal open doors or touch surfaces
- Can also be used as mats and liners; rolls can be cut to desired length
- One time use only
Specifications
Biohazard Wipes | |
22.86 cm | |
20.32 cm | |
Orange backing | |
9 x 8 in. (22.86 x 20.32 cm) | |
Blood tubes, microbiology plates, pipettes, probes, specimen containers, instruments |
9 in. | |
8 in. | |
Heavy | |
Single-use | |
Prevent soak through of blood, body fluids, tissue and chemicals to gloves and surfaces when handling tubes, specimens, plates. |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fisherbrand™ BloodBloc™ Biohazard Wipes, Liners and Rolls > Heavy Absorbency, 8 x 9 in.
Spot an opportunity for improvement?Share a Content Correction