Learn More
AZOWIPE™ Wipes
Bactericidal hard surface disinfectant wipes. AZOWIPE™ Wipes are convenient for use in all clinical environments and are an important part of a robust infection control protocol.
Brand: Vernacare 81160
Description
- Made from a viscose rayon nonwoven material strong and durable in use
- Foil sealed - prevents evaporation in storage
- Suitable for all hard surfaces - one product from a single source
- Contains NO preservative - no smearing or streaking when in use
- Specially designed canister - easy to dispense wipes
- 70% Solution of Isopropyl Alcohol BP
- AZOWIPE - for the disinfection of hard surfaces
- Dimensions: 200mm x 220mm
Specifications
Wipes | |
220 x 200 mm | |
200 Pack |
Product Suggestions
Customers who viewed this item also viewed.
Safety and Handling
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.