Learn More
Fisherbrand™ Combs for Midi-Plus Wide Format Horizontal Gel Units, 1.5mm Thick
Description
- Recommended to scale-up nucleic acid volumes for preparatory techniques
- To maximise sample volume
- Height-adjustable without any requirement for specialist tools or comb holders, to give user full control over well depth and sample-loading volume
- Rigid comb back prevents heat-induced warping
- Reversible loading guides sit directly above each well to provide a convenient loading template for single- and multi-channel pipets
- 12-sample, 25-sample and 50-sample combs are Multichannel pipettor compatible
Specifications
Red | |
Comb | |
1 Each | |
45 ÎĽL |
Midi-Plus Wide Format Horizontal Gel Units | |
25 | |
1.5 mm |
Product Suggestions
Customers who viewed this item also viewed
Videos
-
Fisherbrand and Fisher BioReagents Focus on electrophoresis
Your input is important to us. Please complete this form to provide feedback related to the content on this product.