missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Combs for SUB-GEL Mini Horizontal Gel Electrophoresis Units, 1mm Thick
Description
- Recommended for high resolution gels and publication quality data
- Supplied as standard
- Height-adjustable without any requirement for specialist tools or comb holders, to give user full control over well depth and sample-loading volume
- Rigid comb back prevents heat-induced warping
- Reversible loading guides sit directly above each well to provide a convenient loading template for single- and multi-channel pipets
- 8-sample and 12-sample combs are Multichannel pipettor compatible
Specifications
White | |
Comb | |
1 Each | |
90 μL |
Sub-Gel Mini Horizontal Gel Electrophoresis Unit | |
2 Prep, 2 Marker | |
1 mm |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Videos
-
Fisherbrand and Fisher BioReagents Focus on electrophoresis
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fisherbrand™ Combs for SUB-GEL Mini Horizontal Gel Electrophoresis Units, 1mm Thick > Sample size: 90μL
Spot an opportunity for improvement?Share a Content Correction