Learn More
Fisherbrand™ High-Form Porcelain Crucibles
Alert:
Alert: Heating and cooling rates should not exceed 200°C per hour and maximum use temp of 1150°C
Description
Withstand temperatures up to 1150°C (2102°F)
- Glazed inside and out except for outside bottom surface and rim
Order Info
Covers available separately. See Fisherbrand™ Porcelain Crucible Covers (FB965R through FB965Z).
Specifications
Porcelain | |
1150°C | |
8g | |
24mm | |
Crucible |
Glazed | |
1.8mm | |
5 mL | |
20mm |
Product Suggestions
Customers who viewed this item also viewed.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.