missing translation for 'onlineSavingsMsg'
Learn More
Gallocyanine, Thermo Scientific Chemicals Product Code.: 10236200

Gallocyanine, Thermo Scientific Chemicals

Product Code. 10236200
10 g, Glass bottle
Click to view available options
Quantity:
10 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10236200

Brand: Thermo Scientific Acros 154430100

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1562-85-2
Molecular Formula C15H13ClN2O5
Molecular Weight (g/mol) 336.72
MDL Number MFCD00011926
InChI Key AQSOTOUQTVJNMY-UHFFFAOYSA-N
Synonym gallocyanine, gallocyanin, solid violet, 1-carboxy-7-dimethylamino-3,4-dihydroxyphenoxazin-5-ium chloride, gallocyanine bs, gallocyanine dh, fast violet, anthracene blue swgg, alizarine navy blue at, brilliant chrome blue pShow MoreShow Less
PubChem CID 73801
IUPAC Name 7-(dimethylamino)-4-hydroxy-3-oxophenoxazin-10-ium-1-carboxylic acid;chloride
SMILES CN(C)C1=CC2=C(C=C1)[NH+]=C3C(=CC(=O)C(=C3O2)O)C(=O)O.[Cl-]

Specifications

Color Black to Green
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 10 g
Beilstein 27, 438
Merck Index 15, 4385
Formula Weight 336.72
Physical Form Fine Crystalline Powder
Chemical Name or Material Gallocyanine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0434816 Certificate TypeCertificate of Analysis Date18/04/2024 Catalog Number154430250,154430100
Lot NumberA0148839 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number154430250,154430100
Lot NumberA0152941 Certificate TypeCertificate of Analysis Date06/08/2022 Catalog Number154430250,154430100
compliance-icons
Product Identifier
  • Gallocyanine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gallocyanine, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.