missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ DMEM, high glucose, HEPES, no phenol red
GREENER_CHOICE

DMEM (Dulbecco's Modified Eagle Medium) is a widely used basal medium for supporting the growth of many different mammalian cells.

Brand:  Gibco™ 21063029

17.75 GBP valid until 2025-12-31
Use promo code "24273" to get your promotional price.


 View more versions of this product

Product Code. 11520556

  • £20.23 / 500mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 

Requires : Supplementation
This item is not returnable. View return policy

Description

Description

DMEM (Dulbecco's Modified Eagle Medium) is a widely used basal medium for supporting the growth of many different mammalian cells. Cells successfully cultured in DMEM include primary fibroblasts, neurons, glial cells, HUVECs, and smooth muscle cells, as well as cell lines such as HeLa, 293, Cos-7, and PC-12. We offer a variety of DMEM modifications for a range of cell culture applications. Find the right formulation using the media selector tool.

This DMEM is modified as follows:

With: High Glucose, L-glutamine, HEPES

Without: Sodium Pyruvate, Phenol Red

The complete formulation is available.

Using DMEM

DMEM is unique from other media as it contains 4 times the concentration of amino acids and vitamins than the original Eagle's Minimal Essential Medium. DMEM was originally formulated with low glucose (1 g/L) and sodium pyruvate, but is often used with higher glucose levels, with or without sodium pyruvate. DMEM contains no proteins, lipids, or growth factors. Therefore, DMEM requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). DMEM uses a sodium bicarbonate buffer system (3.7 g/L), and therefore requires a 5–10% CO2 environment to maintain physiological pH.

Order Info

Shipping Condition: Room Temperature

Compliance

DMEM is manufactured at a cGMP-compliant facility located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

HeLa, 293, Cos-7, and PC-12
Animal Origin-free
Liquid
Standard Serum Supplementation
Sterile-filtered
No Phenol Red, No Sodium Pyruvate
cGMP-compliant under the ISO 13485 standard
500 mL
Storage conditions: 2-8° C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture
Primary Fibroblasts, Neurons, Glial Cells, HUVECs, Smooth Muscle Cells
1 X
DMEM (Dulbecco's Modified Eagle Medium)
Sterile-filtered
High Glucose, Glutamine, HEPES
4500 mg/L
Gibco™
Room Temperature
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2508956 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number21063029
Lot Number2579414 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number21063029
Lot Number2660220 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number21063029
Lot Number2756225 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number21063029
Lot Number2666512 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number21063029
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gibco™ DMEM, high glucose, HEPES, no phenol red > Quantity: 500 mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.