missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Liver Cancer Cell Culture Starter Kit, Europe

Provides researchers with a convenient package of reagents for the culture and transfection of cancer cell types frequently used in cancer research.

Brand:  Gibco™ A3518601

Product Code. 16346248

This item is not returnable. View return policy

Description

Description

  • Gibco Cancer Cell Culture Starter kits provide researchers with a convenient package of reagents for the culture and transfection of cancer cell types frequently used in cancer research. Cancer Cell Culture Starter kits are currently available for breast, lung, prostate, and liver cancer cells.
  • Each starter kit includes the following products for cancer cell culture and transfection: Basal media, Fetal bovine serum, Penicillin-streptomycin, TrypLE Express Enzyme, Lipofectamine 3000 Transfection Reagent, Opti-MEM I
  • The Cancer Cell Culture Starter kits contain the most commonly used reagents to help you start your cancer cell culture quickly and achieve successful transfections, based on a review of hundreds of cancer research publications in which the following cell lines were used: Prostate cancer: DU145, PC-3, LNCaP, Lung cancer: A549, NCI-H975, PC-14, NCI-H1299, Breast cancer: MCF-7, MDA-MB-231, T-47D, BT-474, Liver cancer: Hep G2, Hep 3B
  • The Cancer Culture Starter kits also include a suggested protocol for the culture, dissociation (cell passage), and transfection of the most-published cancer cell lines. Achieve peace of mind and quickly initiate your experiments with these starter kits, the components of which may also be purchased separately.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

• DMEM, high glucose, with GlutaMAX Supplement (store at 2–8°C)
• Fetal bovine serum, qualified, One Shot™ format, EU Approved South America origin (store at -5 to -30°C)
• Penicillin-streptomycin (10,000 U/mL) (store at -5 to -30°C)
• TrypLE Express Enzyme (1X), no phenol red (store at room temperature)
• Lipofectamine 3000 Transfection Reagent (store at 2–8°C)
• Opti-MEM I Reduced Serum Medium (store at 2–8°C)

Culture Starter Kit
Kit
1 Kit
Cancer Cells
Sterile-filtered
Mammalian Cell Culture
Approved for shipment at Room Temperature or on Wet or Dry Ice
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gibco™ Liver Cancer Cell Culture Starter Kit, Europe >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.