Learn More
Gilson™ PIPETMAN Neo™ Multichannel Pipets
Offers an excellent compromise between comfort and target precision. Gilson™ PIPETMAN Neo™ Multichannel Pipets are designed to apply equal pressure on each piston to ensure consistent volume levels across all channels.
Brand:  Gilson™ F14401
 View more versions of this product
Product Code. 10773686
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Easy-to-remove tip ejector with autoclavable lower part
  • Internal design applies equal pressure on each piston to ensure consistent volume levels across all channels
  • PVDF handle minimizes the hand-warming effect during pipetting and is resistant to aggressive chemicals
  • Gilson Trident and unique spacer create a stepped ejection, staggering the tip ejection and thus reducing the force required to eject tips
  • Unique tip holder allows for a secure fit of most brands of tips
  • Available in 8- and 12-channel models
Order Info
Trade-in program available. Please contact your Fisher Scientific Sales representative for more details.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Autoclavable
8
Multichannel Pipette
1 Year
P8x20N
Pipetman Neo
2 to 20 μL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products
Special Offers
Special Offers
Product Content Correction
Product Title
Gilson™ PIPETMAN Neo™ Multichannel Pipets > Model P8x20N; 2 - 20μL