Learn More
Hirschmann™ ringcaps™ Disposable Capillary Pipettes
Excellent for working with small samples in the laboratory. Hirschmann Instruments™ ringcaps™ Disposable Capillary Pipettes can be utilized for a wide range of scientific and medical applications.
Brand:  Hirschmann™ 9602144
 View more versions of this product
Product Code. 15559035
  • £136.00 / Pack of 1000
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • With ringmark
  • Conformity certified
  • Manufactured from durable DURAN™ glass
  • Acc. to ISO 7550, CE acc. IVD 98/79 EC
Specifications
Specifications
0.25%
Glass
44.7 μL
Single-use
ISO 7550, CE, IVD 98/79 EC
Ring Mark
Capillary Pipette
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Hirschmann™ ringcaps™ Disposable Capillary Pipettes > Volume: 44.7 uL