Learn More
Poulten Graf™ Glass/PP/PTFE Fortuna™ Optifix™ Solvent Dispenser
Poulten Graf™ Glass/PP/PTFE Fortuna™ Optifix™ Solvent Dispensers have a dispensing cylinder and piston made of glass.
Brand: Poulten Graf™ 101 08133
This item is not returnable.
View return policy
Description
For dispensing of pure organic solvents. Poulten Graf™ Glass/PP/PTFE Fortuna™ Optifix™ Solvent Dispensers have a dispensing cylinder and piston made of glass.
- Manufactured according to DIN EN ISO 9001:2008 using high quality materials
- Solid dispensing piston made of glass
- Cylinder coated with plastic to protect against breakage
- With certificate, individually numbered
- Ergonomic finger protection cap made of PP with volume setting system
- Graduation on the scale is lasered and easy to read
- Fast and easy volume setting with additional fine adjustment
Specifications
Fortuna Optifix Solvent Dispenser | |
1 to 5 mL | |
Each |
PTFE, Polypropylene, Fiberglass | |
For dispensing solvents |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Product Title
Poulten Graf™ Glass/PP/PTFE Fortuna™ Optifix™ Solvent Dispenser > Volume: 1 to 5 mL
Spot an opportunity for improvement?Share a Content Correction