Learn More
Glucosamine, MedChemExpress
MedChemExpress Glucosamine (D-Glucosamine) is an amino sugar and a prominent precursor in the biochemical synthesis of glycosylated proteins and lipids, is used as a dietary supplement. Glucosamine also is a natural constituent of glycosaminoglycans in the cartilage matrix and synovial fluid, which when administered exogenously, exerts pharmacological effects on osteoarthritic cartilage and chondrocytes.
Brand:  Medchem Express HY-B1125
Encompass_Preferred
Product Code. 16478626
  • £216.59 / 100mg
Explore more special offers
Description
Description
Glucosamine (D-Glucosamine) is an amino sugar and a prominent precursor in the biochemical synthesis of glycosylated proteins and lipids, is used as a dietary supplement. Glucosamine also is a natural constituent of glycosaminoglycans in the cartilage matrix and synovial fluid, which when administered exogenously, exerts pharmacological effects on osteoarthritic cartilage and chondrocytes.
Specifications
Specifications
Glucosamine
White
C6H13NO5
D-Glucosamine Chitosamine
O=C[C@H](N)[C@H]([C@@H]([C@@H](CO)O)O)O
179.17
Research
COVID-19-immunoregulation
3416-24-8
Research
100 mg
H2O : 100 mg/mL (558.13 mM; Need ultrasonic) ∣DMSO : 3.33 mg/mL (18.59 mM; Need ultrasonic)
179.17
97.0%
Powder
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Glucosamine, MedChemExpress