Learn More
Glutarimide, 98%, Thermo Scientific Chemicals
Description
Glutarimide acts as an inhibitor of protein synthesis. Further, it is used as a reactant for thionations and biocatalytic asymmetric synthesis of sitagliptin production. It is also employed in the generation of beta-adrenoceptor ligands, enantioselective synthesis of securinega alkaloids and alfa-fluoro-alfa amino amides. In addition to this, it is used in intramolecular amidocyclopropanation reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Glutarimide acts as an inhibitor of protein synthesis. Further, it is used as a reactant for thionations and biocatalytic asymmetric synthesis of sitagliptin production. It is also employed in the generation of beta-adrenoceptor ligands, enantioselective synthesis of securinega alkaloids and alfa-fluoro-alfa amino amides. In addition to this, it is used in intramolecular amidocyclopropanation reactions.
Solubility
Soluble in water, hot ethanol and boiling benzene. Insoluble in ether.
Notes
Incompatible with strong oxidizing agents.

Specifications
Specifications
Melting Point | 153°C to 157°C |
Odor | Nearly Odorless |
Quantity | 25 g |
Beilstein | 110052 |
Solubility Information | Soluble in water,hot ethanol and boiling benzene. Insoluble in ether. |
Formula Weight | 113.12 |
Percent Purity | 98% |
Chemical Name or Material | Glutarimide |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
10223917 | Certificate of Analysis | 30/08/2024 | L00968.22,L00968.14,L00968.06 |
10205459 | Certificate of Analysis | 06/07/2023 | L00968,L00968.22,L00968.14,L00968.06 |
10113901 | Certificate of Analysis | 08/12/2021 | L00968.22,L00968.14,L00968.06 |
10034035 | Certificate of Analysis | 08/12/2021 | L00968.22,L00968.14,L00968.06 |
10203792 | Certificate of Analysis | 07/12/2021 | L00968.22,L00968.14,L00968.06 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.