missing translation for 'onlineSavingsMsg'
Learn More
Glutarimide, 98%, Thermo Scientific Chemicals Product Code.: 11394394

Glutarimide, 98%, Thermo Scientific Chemicals

Product Code. 11394394
5 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11394394

Brand: Thermo Scientific Alfa Aesar L00968.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Glutarimide acts as an inhibitor of protein synthesis. Further, it is used as a reactant for thionations and biocatalytic asymmetric synthesis of sitagliptin production. It is also employed in the generation of beta-adrenoceptor ligands, enantioselective synthesis of securinega alkaloids and alfa-fluoro-alfa amino amides. In addition to this, it is used in intramolecular amidocyclopropanation reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glutarimide acts as an inhibitor of protein synthesis. Further, it is used as a reactant for thionations and biocatalytic asymmetric synthesis of sitagliptin production. It is also employed in the generation of beta-adrenoceptor ligands, enantioselective synthesis of securinega alkaloids and alfa-fluoro-alfa amino amides. In addition to this, it is used in intramolecular amidocyclopropanation reactions.

Solubility
Soluble in water, hot ethanol and boiling benzene. Insoluble in ether.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1121-89-7
Molecular Formula C5H7NO2
Molecular Weight (g/mol) 113.12
MDL Number MFCD00006670
InChI Key KNCYXPMJDCCGSJ-UHFFFAOYSA-N
Synonym glutarimide, 2,6-piperidinedione, 2,6-diketopiperidine, 2,6-dioxopiperidine, acetylglycinamide impurity b, piperidin-2,6-dione, acmc-2099da, 5-21-09-00557 beilstein handbook reference, 2,6-piperidinedione 9ciShow MoreShow Less
PubChem CID 70726
ChEBI CHEBI:5435
IUPAC Name piperidine-2,6-dione
SMILES O=C1CCCC(=O)N1

Specifications

Melting Point 153°C to 157°C
Odor Nearly Odorless
Quantity 5 g
Beilstein 110052
Solubility Information Soluble in water,hot ethanol and boiling benzene. Insoluble in ether.
Formula Weight 113.12
Percent Purity 98%
Chemical Name or Material Glutarimide

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10223917 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberL00968.22,L00968.14,L00968.06
Lot Number10205459 Certificate TypeCertificate of Analysis Date06/07/2023 Catalog NumberL00968,L00968.22,L00968.14,L00968.06
Lot Number10113901 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL00968.22,L00968.14,L00968.06
Lot Number10034035 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL00968.22,L00968.14,L00968.06
Lot Number10203792 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL00968.22,L00968.14,L00968.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Glutarimide, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.