missing translation for 'onlineSavingsMsg'
Learn More

Glutathione Reduced, Fisher BioReagents

£43.80 - £327.00

Chemical Identifiers

CAS 70-18-8
Molecular Formula C10H17N3O6S
Molecular Weight (g/mol) 307.321
InChI Key RWSXRVCMGQZWBV-WDSKDSINSA-N
Synonym glutathione, l-glutathione, glutathion, glutathione-sh, glutinal, isethion, tathion, reduced glutathione, deltathione, neuthion
PubChem CID 124886
ChEBI CHEBI:16856
IUPAC Name (2S)-2-amino-5-[[(2R)-1-(carboxymethylamino)-1-oxo-3-sulfanylpropan-2-yl]amino]-5-oxopentanoic acid
SMILES C(CC(=O)NC(CS)C(=O)NCC(=O)O)C(C(=O)O)N
View More Specs

Products 6
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10071213
View Documents
Fisher BioReagents
BP2521-1
1 g Amber Glass This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10562785
View Documents
Fisher BioReagents
BP2521-5
5 g Amber Glass This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10204873
View Documents
Fisher BioReagents
BP2521-10
10 g Amber Glass This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10482475
View Documents
Fisher BioReagents
BP2521-25
25 g Amber Glass This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11483074
View Documents
Fisher BioReagents
BP2521-50
50 g Amber Glass This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10566394
View Documents
Fisher BioReagents
BP2521-100
100 g Amber Glass This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Glutathione is a useful tripeptide involved in many aspects of metabolism, including transport of γ-glutamyl amino acids and reductive cleavage of disulfide bonds.
Specifications

Chemical Identifiers

70-18-8
307.321
glutathione, l-glutathione, glutathion, glutathione-sh, glutinal, isethion, tathion, reduced glutathione, deltathione, neuthion
CHEBI:16856
C(CC(=O)NC(CS)C(=O)NCC(=O)O)C(C(=O)O)N
C10H17N3O6S
RWSXRVCMGQZWBV-WDSKDSINSA-N
124886
(2S)-2-amino-5-[[(2R)-1-(carboxymethylamino)-1-oxo-3-sulfanylpropan-2-yl]amino]-5-oxopentanoic acid

Specifications

70-18-8
White
0.01% max.
≥98 %
15, 4511
glutathione, l-glutathione, glutathion, glutathione-sh, glutinal, isethion, tathion, reduced glutathione, deltathione, neuthion
C(CC(=O)NC(CS)C(=O)NCC(=O)O)C(C(=O)O)N
307.321
CHEBI:16856
≥98%
Glutathione Reduced
182°C
3
Report %
C10H17N3O6S
-21 to -11° (+ or -)
RWSXRVCMGQZWBV-WDSKDSINSA-N
(2S)-2-amino-5-[[(2R)-1-(carboxymethylamino)-1-oxo-3-sulfanylpropan-2-yl]amino]-5-oxopentanoic acid
124886
307.3
Powder or Solid
Videos
Safety and Handling

Safety and Handling

Recommended Storage : 4°C

Product Identifier
  • Glutathione reduced

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.