missing translation for 'onlineSavingsMsg'
Learn More
Glycerol, ACS, 99.5+%, Thermo Scientific Chemicals Product Code.: 11405190

Glycerol, ACS, 99.5+%, Thermo Scientific Chemicals

Product Code. 11405190
500 mL
Click to view available options
Quantity:
500 mL
1 L
4 L
4 x 1 L
This item is not returnable. View return policy

Product Code. 11405190

Brand: Thermo Scientific Alfa Aesar 036646.AP

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Glycerol is widely used in cosmetics, especially for production of soaps and lotions, solvents and emulsifiers. It may also be found in inks, adhesives and pharmaceuticals. It plays an important role in the manufacture of plastics and resins. Glycerol bonded with silica gel plays an important role for effective removal of traces of Co(II) from aqueous solutions. Also useful for optical clearing of gastric tissue.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-81-5
Molecular Formula C3H8O3
Molecular Weight (g/mol) 92.09
MDL Number MFCD00004722
InChI Key PEDCQBHIVMGVHV-UHFFFAOYSA-N
Synonym glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglynShow MoreShow Less
PubChem CID 753
ChEBI CHEBI:17754
IUPAC Name propane-1,2,3-triol
SMILES OCC(O)CO

Specifications

Melting Point 18°C
Density 1.26 g/cm3
Boiling Point 182°C (20 mm)
Flash Point 160°C (320°F)
Ignition Point 400°C (752°F)
Assay Percent Range >99.5%
Refractive Index 1.474
Quantity 500 mL
Beilstein 635685
Sensitivity Hygroscopic
Solubility Information Miscible in water,methanol,ethanol and 2-propanol. Immiscible in ether,benzene and chloroform.
Formula Weight 92.09
Vapor Pressure <0.1 hPa (0mm Hg) at 20°C
Grade ACS
Physical Form Viscous Liquid
Chemical Name or Material Glycerol
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberP05L015 Certificate TypeCertificate of Analysis Date07/03/2025 Catalog Number036646.M1,036646.AP,036646.K7,036646.K2
Lot NumberU16K004 Certificate TypeCertificate of Analysis Date28/08/2024 Catalog Number036646.M1,036646.AP,036646.K7,036646.K2
Lot NumberY03I015 Certificate TypeCertificate of Analysis Date11/08/2023 Catalog Number036646.M1,036646.AP,036646.K7,036646.K2
Lot NumberZ19I009 Certificate TypeCertificate of Analysis Date11/01/2023 Catalog Number036646.M1,036646.AP,036646.K7,036646.K2
Lot NumberY03I016 Certificate TypeCertificate of Analysis Date09/11/2022 Catalog Number036646.M1,036646.AP,036646.K7,036646.K2

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Glycerol, ACS, 99.5+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.