missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
Specifications
Density | 1.2580 to 1.2644g/mL at 20°C, ≥1.257g/mL at 25°C |
Flash Point | 160°C (320°F) |
Ignition Residue | ≤0.01% (as SO4) |
Ignition Point | 698°F |
Linear Formula | HOCH2CH(OH)CH2OH |
Quantity | 1 L |
UN Number | NONH for all modes of transport |
Beilstein | 635685 |
Vapor Density | 3.1 (vs air) |
Formula Weight | 92.09g/mol |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 9
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Safety and Handling
Product Identifier
- Glycerol
Special Offers
-
Multiple Manufacturers
Chemical Extravaganza
Shop now and save up to 97% on top-quality products for your lab needs.Promo Code: 25908Expires: 2025-03-31
Days
0Hours
11Minutes
24Seconds
23
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Glycerol, puriss., Honeywell >
Spot an opportunity for improvement?Share a Content Correction