missing translation for 'onlineSavingsMsg'
Learn More
Glycerol, ultrapure, 99.5+%, Thermo Scientific Chemicals Product Code.: 15456539

Glycerol, ultrapure, 99.5+%, Thermo Scientific Chemicals

Product Code. 15456539
100 mL
Click to view available options
Quantity:
100 mL
250 mL
500 mL
This item is not returnable. View return policy

Product Code. 15456539

Brand: Thermo Scientific Alfa Aesar J64719.AE

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used in sample preparation and gel formation for polyacrylamide gel electrophoresis. Glycerol (5-10%) increases the density of a sample so that the sample will layer at the bottom of a gel’s sample well.

Glycerol, ultrapure is used in sample preparation and gel formation for polyacrylamide gel electrophoresis. Glycerol (5-10%) increases the density of a sample so that the sample will layer at the bottom of a gel's sample well. Glycerol is also used to aid in casting gradient gels and as a protein stabilizer and storage buffer component, glycerol is used as a solvent, plasticizer and humectant. Additional uses include as an aid in casting gradient gels, a protein stabilizer and storage buffer component.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-81-5
Molecular Formula C3H8O3
Molecular Weight (g/mol) 92.09
MDL Number MFCD00004722
InChI Key PEDCQBHIVMGVHV-UHFFFAOYSA-N
Synonym glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglynShow MoreShow Less
PubChem CID 753
ChEBI CHEBI:17754
IUPAC Name propane-1,2,3-triol
SMILES OCC(O)CO

Specifications

Melting Point 18°C
Density 1.26
Boiling Point 182°C (20 mmHg)
Flash Point 160°C (320°F)
Packaging Plastic bottle
Linear Formula HOCH2CH(OH)CH2OH
Refractive Index 1.474
Quantity 100 mL
Beilstein 635685
Sensitivity Hygroscopic
Specific Gravity 1.26
Solubility Information Miscible with water
Formula Weight 92.09
Percent Purity ≥99.5%
Physical Form Viscous Liquid
Chemical Name or Material Glycerol
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberY17I025 Certificate TypeCertificate of Analysis Date24/10/2023 Catalog NumberJ64719.AK,J64719.AP,J64719.AE
Lot NumberU30J031 Certificate TypeCertificate of Analysis Date21/09/2023 Catalog NumberJ64719.AK,J64719.AP,J64719.AE
Lot NumberU30J032 Certificate TypeCertificate of Analysis Date21/09/2023 Catalog NumberJ64719.AK,J64719.AP,J64719.AE
Lot NumberY14I013 Certificate TypeCertificate of Analysis Date29/11/2022 Catalog NumberJ64719.AK,J64719.AP,J64719.AE
Lot NumberX12I020 Certificate TypeCertificate of Analysis Date29/11/2022 Catalog NumberJ64719.AK,J64719.AP,J64719.AE

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Glycerol, ultrapure, 99.5+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.