missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Product Code | Brand | Dimensions (Dia. x H) | Well Count | Vent | Sterility | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (Dia. x H) | Well Count | Vent | Sterility | Price | Quantity & Availability | ||||||
11861583
|
Corning™
BP93B-16 |
90 x 15 mm | 1 | 3 | Sterile | N/A | N/A | N/A | |||||
12374079
|
Corning™
BI93D-01 |
90 x 15 mm | 2 | 3 | Aseptic | N/A | N/A | N/A | |||||
11812532
|
Corning™
BP53-01 |
60 x 15 mm | 1 | 6 | Non-sterile | N/A | N/A | N/A | |||||
11768573
|
Corning™
BP53-02 |
60 x 15 mm | 1 | 6 | Sterile | N/A | N/A | N/A | |||||
11831553
|
Corning™
BP93B-15 |
90 x 15 mm | 1 | 3 | Aseptic | N/A | N/A | N/A | |||||
12364069
![]() ![]() |
Corning™
BB64-01 |
65 x 14.5 mm | 1 | 6 | Sterile |
£486.00
Pack of 720 |
Estimated Shipment: 24-06-2025
Log in to see stock. |
||||||
12397487
|
Corning™
SB93-03 |
90 x 15 mm | 1 | 3 | Sterile | N/A | N/A | N/A | |||||
11798563
![]() |
Corning™
BP124-04 |
120.5 x 15.8 mm | 1 | 4 | Sterile or Aseptic |
£229.00
Pack of 252 |
In Stock
Log in to see stock. ![]() |
||||||
11728573
![]() ![]() |
Corning™
BP63-10 |
65 x 14.5 mm | 1 | 3 | Sterile |
£208.00
Pack of 720 |
Estimated Shipment: 14-07-2025
Log in to see stock. |
||||||
15488784
![]() |
Corning™
BP93B-102 |
90 x 14.2 mm | 1 | 4 | Sterile |
£201.00
Pack of 825 |
In Stock
Log in to see stock. |
||||||
15498784
![]() |
Corning™
BP93D-102 |
90 x 14.2 mm | 1 | 3 | Sterile |
£107.41
Pack of 825 |
Estimated Shipment: 02-09-2025
Log in to see stock. |
||||||
15446254
|
Corning™
BP53-03 |
55 x 14.2 mm | 1 | 6 | Sterile |
£449.00
Pack of 1620 |
In Stock
Log in to see stock. ![]() |
||||||
Description
Optimized for automation. Gosselin™ Petri Dishes are manufactured from crystal polystyrene.
- Crystal polystyrene
- Optimal flatness
- Manufactured in class 100
- Easy opening
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Corning™ Petri Dishes