missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Halt™ Phosphatase Inhibitor Single-Use Cocktail

Stabilize serine, threonine and tyrosine phosphorylation of proteins in cells and tissues with this cocktail of phosphatase inhibitors.

Brand:  Thermo Scientific™ 78428

288.80 GBP valid until 2025-05-31
Use promo code "25977" to get your promotional price.



 View more versions of this product

Product Code. 11833955

  • £311.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

Preserve phosphorylation of proteins in cells and tissues with Thermo Scientific™ Halt Phosphatase Inhibitor Cocktail.

  • Halt Phosphatase Inhibitor Cocktail protects phosphoproteins from serine/threonine phosphatases, as well as protein tyrosine phosphatases (PTPs) during cell lysis
  • The cocktail contains a mixture of four inhibitors of broad specificity including sodium fluoride, sodium orthovanadate, sodium pyrophosphate, and beta-glycerophosphate
  • Conveniently packaged in a 100X format, the cocktail is stable at 4°C and is suitable for use with any cell or tissue extract
  • The cocktail preserves protein phosphorylation in immunoprecipitation and kinase assays
  • Phosphorylation and dephosphorylation comprise a molecular on/off switch that regulates many key biological pathways within the cell including signal transduction, cell division, and apoptosis
  • To effectively study phosphorylation activation states, one must inactivate phosphatases to prevent their uncontrolled activity upon cell lysis

Additional Features

  • Devoid of protease inhibitors – To protect against both proteases and phosphatases, use Halt Protease and Phosphatase Inhibitor Cocktail (78420)
  • Dialyzable – Either dialyze or desalt sample to effectively remove inhibitors from cell extracts before performing procedures that are discovered to be incompatible with cocktail components
  • Supplied as a 100X concentrate – effective when used at a final concentration of 1X; however, samples that contain particularly high levels of phosphatase activity may require a final concentration of 2-3X
  • Compatibility – compatible with most detergent-based cell lysis reagents, including all Thermo Scientific Pierce Cell Lysis Solutions
  • All-in-one format – inhibits both serine-threonine phosphatases and protein tyrosine phosphatases
  • Compatible with popular protein assays – works with standard protein assays, including BCA and coomassie (Bradford) assays
  • Convenient storage – refrigerator storage (2 to 8°C) saves valuable space in the freezer and allows for immediate use
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Phosphatase Inhibitor Cocktail
Liquid
Phosphatase Inhibitor
24 x 100 μL
Upon receipt store product at 4°C. Do not freeze.
Halt™
Phosphatase Inhibitor
Product Suggestions

Product Suggestions

Videos
Safety and Handling
Show More
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZL397936 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number78428
Lot NumberZI394882 Certificate TypeCertificate of Analysis Date15/10/2024 Catalog Number78428
Lot NumberZF391250 Certificate TypeCertificate of Analysis Date17/07/2024 Catalog Number78428
Lot NumberZC389937 Certificate TypeCertificate of Analysis Date04/04/2024 Catalog Number78428
Lot NumberZA386958 Certificate TypeCertificate of Analysis Date10/02/2024 Catalog Number78428
Special Offers

Special Offers

  • Multiple Manufacturers
    Advance your proteomics research with essential tools
    Promo Code: 25977

    Expires2025-05-31

    Days

    43

    Hours

    16

    Minutes

    27

    Seconds

    21
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Halt™ Phosphatase Inhibitor Single-Use Cocktail > 24 x 100 μL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.