missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Halt™ Phosphatase Inhibitor Single-Use Cocktail
Stabilize serine, threonine and tyrosine phosphorylation of proteins in cells and tissues with this cocktail of phosphatase inhibitors.
£201.00 - £796.00
Specifications
Product Type | Phosphatase Inhibitor Cocktail |
---|---|
Content And Storage | Upon receipt store product at 4°C. Do not freeze. |
Form | Liquid |
Product Line | Halt™ |
Reagent Type | Phosphatase Inhibitor |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10668304
|
Thermo Scientific™
78420 |
1 mL |
£201.00
1mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11833955
|
Thermo Scientific™
78428 |
24 x 100 μL |
£311.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11814101
|
Thermo Scientific™
78426 |
5 x 1 mL |
£588.00
5mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11824101
|
Thermo Scientific™
78427 |
10 mL |
£796.00
10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Preserve phosphorylation of proteins in cells and tissues with Thermo Scientific™ Halt Phosphatase Inhibitor Cocktail.
- Halt Phosphatase Inhibitor Cocktail protects phosphoproteins from serine/threonine phosphatases, as well as protein tyrosine phosphatases (PTPs) during cell lysis
- The cocktail contains a mixture of four inhibitors of broad specificity including sodium fluoride, sodium orthovanadate, sodium pyrophosphate, and beta-glycerophosphate
- Conveniently packaged in a 100X format, the cocktail is stable at 4°C and is suitable for use with any cell or tissue extract
- The cocktail preserves protein phosphorylation in immunoprecipitation and kinase assays
- Phosphorylation and dephosphorylation comprise a molecular on/off switch that regulates many key biological pathways within the cell including signal transduction, cell division, and apoptosis
- To effectively study phosphorylation activation states, one must inactivate phosphatases to prevent their uncontrolled activity upon cell lysis
Additional Features
- Devoid of protease inhibitors – To protect against both proteases and phosphatases, use Halt Protease and Phosphatase Inhibitor Cocktail (78420)
- Dialyzable – Either dialyze or desalt sample to effectively remove inhibitors from cell extracts before performing procedures that are discovered to be incompatible with cocktail components
- Supplied as a 100X concentrate – effective when used at a final concentration of 1X; however, samples that contain particularly high levels of phosphatase activity may require a final concentration of 2-3X
- Compatibility – compatible with most detergent-based cell lysis reagents, including all Thermo Scientific Pierce Cell Lysis Solutions
- All-in-one format – inhibits both serine-threonine phosphatases and protein tyrosine phosphatases
- Compatible with popular protein assays – works with standard protein assays, including BCA and coomassie (Bradford) assays
- Convenient storage – refrigerator storage (2 to 8°C) saves valuable space in the freezer and allows for immediate use
Specifications
Phosphatase Inhibitor Cocktail | |
Liquid | |
Phosphatase Inhibitor |
Upon receipt store product at 4°C. Do not freeze. | |
Halt™ | |
Phosphatase Inhibitor |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Halt Phosphatase Inhibitor Cocktail
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
Hazard Statement
- H302-Harmful if swallowed.
Precautionary Statement
- P270-Do not eat, drink or smoke when using this product.
- P264-Wash thoroughly after handling.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P330-Rinse mouth.
- P501-Dispose of contents/container to .
Supplemental information
- MIXTURE LIST-Contains : sodium fluoride, trisodium tetraoxovanadate
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thermo Scientific™ Halt™ Phosphatase Inhibitor Single-Use Cocktail