missing translation for 'onlineSavingsMsg'
Learn More
Learn More
asecos™ Hazardous Substances Expandable Rack
The Hazardous Substances Rack provides storage of water pollutants with a steel sump on each storage level.
Brand: asecos™ 27836
This item is not returnable.
View return policy
Description
- Galvanised rack frames
- Sturdy construction due to reinforcement cross bars
- Storage levels in 25 mm increments, height-adjustable
- Load capacity per storage shelf: 200 kg (evenly distributed)
- Flat-pack delivery with assembly instructions
- 4 painted sumps made of steel as storage levels
Specifications
10.6 gal. | |
Steel | |
51.65 in. | |
51.65 in. | |
78.74 in. | |
25.08 in. | |
246.92 lb. |
40 L | |
Drum Rack | |
1312.00 mm | |
1312.00 mm | |
2000.00 mm | |
637.00 mm | |
112 kg |
Product Suggestions
Customers who viewed this item also viewed
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
asecos™ Hazardous Substances Expandable Rack > Capacity: 40 L
Spot an opportunity for improvement?Share a Content Correction