missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HEPES, >98%, MP Biomedicals™
£27.70 - £517.00
Chemical Identifiers
CAS | 7365-45-9 |
---|---|
Molecular Formula | C8H18N2O4S |
Molecular Weight (g/mol) | 238.30 |
MDL Number | MFCD00006158 |
InChI Key | JKMHFZQWWAIEOD-UHFFFAOYSA-N |
Synonym | 4-(2-Hydroxyethyl)piperazine-1-ethanesulfonic acid sodium salt, N-(2-Hydroxyethyl)piperazine-N?-(2-ethanesulfonic acid) sodium salt |
PubChem CID | 23831 |
ChEBI | CHEBI:42334 |
SMILES | OCCN1CCN(CCS(O)(=O)=O)CC1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11400832
|
MP Biomedicals
0219454910 |
10 g |
£27.70
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11410832
|
MP Biomedicals
0219454925 |
25 g |
£38.60
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11420832
|
MP Biomedicals
0219454950 |
50 g |
£62.95
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11430832
|
MP Biomedicals
0219454980 |
100 g |
£93.55
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11440832
|
MP Biomedicals
0219454983 |
250 g |
£182.00
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11450832
|
MP Biomedicals
0219454990 |
500 g |
£388.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11460832
|
MP Biomedicals
0219454991 |
1 kg |
£517.00
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
7365-45-9 | |
238.30 | |
JKMHFZQWWAIEOD-UHFFFAOYSA-N | |
23831 | |
OCCN1CCN(CCS(O)(=O)=O)CC1 |
C8H18N2O4S | |
MFCD00006158 | |
4-(2-Hydroxyethyl)piperazine-1-ethanesulfonic acid sodium salt, N-(2-Hydroxyethyl)piperazine-N?-(2-ethanesulfonic acid) sodium salt | |
CHEBI:42334 |
Specifications
7365-45-9 | |
Powder | |
MFCD00006158 | |
JKMHFZQWWAIEOD-UHFFFAOYSA-N | |
2-[4-(2-hydroxyethyl)piperazin-1-yl]ethanesulfonic acid | |
23831 | |
238.3 | |
HEPES |
White | |
C8H18N2O4S | |
4-(2-Hydroxyethyl)piperazine-1-ethanesulfonic acid sodium salt, N-(2-Hydroxyethyl)piperazine-N?-(2-ethanesulfonic acid) sodium salt | |
OCCN1CCN(CCS(O)(=O)=O)CC1 | |
238.30 | |
CHEBI:42334 | |
>98% |
Sicherheit und Handhabung
missing translation for 'einecsNumber' : 230-907-9
Recommended Storage : Store at Room Temperature (15 to 30°C). Store Desiccated.