missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Herasafe™ KS Options and Accessories

Worksurfaces, exhaust transitions and baskets

Brand:  Thermo Scientific™ 50073671

 View more versions of this product

Product Code. 10134173

  • £802.67 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Worksurfaces, exhaust transitions and baskets

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Indented Worksurface Module 1500 mm Wide (316SS)
Indented Worksurface Module
1500 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Herasafe™ KS Options and Accessories > Indented worksurface module 1500mm wide (316SS)

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.