missing translation for 'onlineSavingsMsg'
Learn More
Hexamethylenetetramine, 99+%, Thermo Scientific Chemicals Product Code.: 11423437

Hexamethylenetetramine, 99+%, Thermo Scientific Chemicals

Product Code. 11423437
1000 g
missing translation for 'orderingAttributeHoverText'
Quantity:
250 g
1000 g
5000 g
This item is not returnable. View return policy

Product Code. 11423437

Brand: Thermo Scientific Alfa Aesar A17213.0B

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Hexamethylenetetramine is used as a formylation agent in the synthesis and as a precipitating agent. It is also used in the production of phenolic resins and its moulding compounds, which finds application as binders in fire proof materials, brake and clutch linings. It is involved in the preparation of 5-tert-butyl-2-hydroxy-isophthalaldehyde. It is useful for the treatment of urinary tract infection. Further, it is used in the Duff reaction, Sommelet reaction and in the Delepine reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hexamethylenetetramine is used as a formylation agent in the synthesis and as a precipitating agent. It is also used in the production of phenolic resins and its moulding compounds, which finds application as binders in fire proof materials, brake and clutch linings. It is involved in the preparation of 5-tert-butyl-2-hydroxy-isophthalaldehyde. It is useful for the treatment of urinary tract infection. Further, it is used in the Duff reaction, Sommelet reaction and in the Delepine reaction.

Solubility
Soluble in water, ethanol, chloroform and acetone.

Notes
Hygroscopic. Incompatible with strong oxidizing agents and strong acids. Avoid conditions like flames, sparks and moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-97-0
Molecular Formula C6H12N4
Molecular Weight (g/mol) 140.19
MDL Number MFCD00006895
InChI Key VKYKSIONXSXAKP-UHFFFAOYSA-N
Synonym methenamine, hexamethylenetetramine, hexamine, urotropine, 1,3,5,7-tetraazaadamantane, hexamethylenamine, aminoform, methenamin, urotropin, formamineShow MoreShow Less
PubChem CID 4101
ChEBI CHEBI:6824
SMILES C1N2CN3CN1CN(C2)C3

Specifications

Melting Point ∼280°C (sublimation)
Density 1.33
Flash Point 250°C (482°F)
Quantity 1000 g
UN Number UN1328
Beilstein 2018
Sensitivity Hygroscopic
Merck Index 14,5966
Solubility Information Soluble in water,ethanol,chloroform and acetone.
Formula Weight 140.19
Percent Purity ≥99%
Chemical Name or Material Hexamethylenetetramine
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10252638 Certificate TypeCertificate of Analysis Date05/10/2024 Catalog NumberA17213.30,A17213.0I,A17213.0B
Lot Number10247881 Certificate TypeCertificate of Analysis Date05/09/2023 Catalog NumberA17213.30,A17213.0I,A17213.0B
Lot Number10238069 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA17213.30,A17213.0I,A17213.0B
Lot Number10239366 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA17213.30,A17213.0I,A17213.0B
Lot Number10238163 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA17213.30,A17213.0I,A17213.0B
compliance-icons compliance-icons
Product Identifier
  • Hexamethylenetetramine
Signal Word
  • Warning
Hazard Category
  • Flammable solid Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H228-Flammable solid.
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P240-Ground/bond container and receiving equipment.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P501-Dispose of contents/container to in accordance with local/regional/national/international regulations..

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Hexamethylenetetramine, 99+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.