missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ High Temperature Autoclavable Bags

Manufactured from polypropylene. Fisherbrand™ High Temperature Autoclave Bags can withstand temperatures up to 134°C

Brand:  Fisherbrand™ SA21-01

350.81 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 11850482

  • £483.00 / Pack of 500
Estimated Shipment: 12-09-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Polypropylene construction
  • 40μm thickness
  • With printed biohazard symbol
  • Supplied in bulk packs of 500
  • Suitable for temperatures of up to 134°C
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Autoclavable
Natural
Clinical, dairy, environment, industries, microbiology
40 μm
780 mm
21 L
Single-use
Polypropylene
134°C
400 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates
Special Offers

Special Offers

  • Multiple Manufacturers
    Use promotion code 24138 to get amazing savings on products for your analysis
    Promo Code: 24138

    Expires2025-04-30

    Days

    6

    Hours

    0

    Minutes

    5

    Seconds

    5
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisherbrand™ High Temperature Autoclavable Bags > Capacity: 21L

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.