Learn More
Thermo Scientific™ HMDS (Hexamethyldisilazane)
Greatly extends the practical range of GC, improving chromatographic results in the silylation of sugars and related substances.
Brand: Thermo Scientific™ TS-84770
285.79 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Thermo Scientific™ HMDS (Hexamethyldisilazane) greatly extends the practical range of GC, improving chromatographic results in the silylation of sugars and related substances.
- Suitable for deactivating and coating chromatographic supports
- Monofunctional, making polymerization not possible and eliminating surface moisture

Specifications
Deactivating and Coating Chromatographic Supports | |
98 to 100% | |
Room Temperature | |
Hexamethyldisilazane |
Corrosive, Flammable | |
25 g | |
25 g |
Product Suggestions
Customers who viewed this item also viewed.
Safety and Handling
- HMDS, 25 g. Hexamethyldisilazane
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Acute toxicity Category 3
- Skin corrosion/irritation Category 2
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H302-Harmful if swallowed.
- H311-Toxic in contact with skin.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H331-Toxic if inhaled.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
- P240-Ground/bond container and receiving equipment.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P242-Use only non-sparking tools.
- P370+P378-In case of fire: Use to extinguish.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P330-Rinse mouth.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P501-Dispose of contents/container to .
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.