missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Honeywell™ Plug
Click to view available options
Noise Level: 33dB
Description
Order Info
dimProductType
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Specifications
Quantity | 500/Pk. |
Noise Reduction Rating (NRR) | 33 dB |
Product Type | Plug |
Product Suggestions
Customers who viewed this item also viewed
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Honeywell™ Plug >
Spot an opportunity for improvement?Share a Content Correction