Learn More
Sperian™ Honeywell™ POLYSAFE™ Clear Uncoated Safety Glasses Product Code.: 11807191
Add to basket
Sperian™ Honeywell™ POLYSAFE™ Clear Uncoated Safety Glasses
Product Code. 11807191
Clear
Click to view available options
Frame Color:
Clear
Product Code. 11807191
Brand: Sperian™ 1002550
Add to basket
Add to basket
For general use and protection against impact up to 45m/s. Sperian™ Honeywell™ POLYSAFE™ Clear Uncoated Safety Glasses fit comfortably over prescription glasses.
  • Good for mechanical work involving grinding, turning and milling
  • Wraparound spectacles for clear and distortion-free vision
  • Indirect vents help prevent fogging
  • Perforated temples to allow the addition of a safety cord
  • Provides 99.9% UV protection
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EN 166 1F, EN 170 and 2-1.2
Lens Tint Clear
Product Type Eye and Face Protection Spectacles
For Use With (Application) Fits over prescription Glasses, General use and protection against impact (45m/s), Mechanic work: Grinding, turning, milling
Frame Color Clear
No. per Pack 10/Pk.
Lens Material Polycarbonate
Product Content Correction
Product Title
Sperian™ Honeywell™ POLYSAFE™ Clear Uncoated Safety Glasses >